Learn More
L-Tyrosine-d4, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | L-Tyrosine-d4 |
| CAS | 62595-14-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C9 2H4 H7 N O3 |
| Quantity | 100 mg |
| Synonym | L-Tyrosine-2,3,5,6-d4, L-Tyrosine-2',3',5',6'-2H4, (2S)-2-Amino-3-(4-hydroxyphenyl-2,3,5,6-d4)propanoic acid, 4-Hydroxyphenyl-d4-L-alanine, β-(p-Hydroxyphenyl)alanine-d4, L-Tyrosine-d4, L-Tyrosine-D4 (4-hydroxyphenyl-2,3,5,6-D4) |
| InChI Formula | InChI=1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1/i1D,2D,3D,4D |
| SMILES | [2H]c1c([2H])c(C[C@H](N)C(=O)O)c([2H])c([2H])c1O |
| IUPAC Name | (2S)-2-amino-3-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)propanoic acid |
| Molecular Weight (g/mol) | 185.21 |
| Show More |