missing translation for 'onlineSavingsMsg'
Learn More

Epsilon-Caprolactam-d10, TRC

Product Code. 30338676
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30338676

Brand: Toronto Research Chemicals TRCC175667100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material eta-Caprolactam-d10
CAS 169297-53-4
Molecular Formula C6HD10NO
Quantity 100 mg
Synonym Hexahydro-3,4,5,6,7-d5-2H-azepin-2-one-3,4,5,6,7-d5
InChI Formula InChI=1S/C6H11NO/c8-6-4-2-1-3-5-7-6/h1-5H2,(H,7,8)/i1D2,2D2,3D2,4D2,5D2
SMILES [2H]C1([2H])NC(=O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC Name 3,3,4,4,5,5,6,6,7,7-decadeuterioazepan-2-one
Molecular Weight (g/mol) 123.22
Formula Weight 123.1468
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.