missing translation for 'onlineSavingsMsg'
Learn More

Deschloro Cetirizine Dihydrochloride, TRC

Product Code. 17913335
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913335 100 mg 100mg
30007415 50 mg 50mg
17972175 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17913335 Supplier Toronto Research Chemicals Supplier No. TRCD288885100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83881-54-3
Molecular Formula C21 H26 N2 O3 . 2 Cl H
Molecular Weight (g/mol) 427.36
Synonym [2-[4-(Diphenylmethyl)piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride, Cetirizine Dihydrochloride Imp. F (EP) as Dihydrochloride, Acetic acid, 2-[2-[4-(diphenylmethyl)-1-piperazinyl]ethoxy]-, hydrochloride (1:2), Acetic acid, [2-[4-(diphenylmethyl)-1-piperazinyl]ethoxy]-, dihydrochloride (9CI)
IUPAC Name 2-[2-(4-benzhydrylpiperazin-1-yl)ethoxy]acetic acid;dihydrochloride
SMILES Cl.Cl.OC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccccc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H26N2O3.2ClH/c24-20(25)17-26-16-15-22-11-13-23(14-12-22)21(18-7-3-1-4-8-18)19-9-5-2-6-10-19;;/h1-10,21H,11-17H2,(H,24,25);2*1H
Formula Weight 426.1477
Percent Purity >95%
Chemical Name or Material (2-[4-(Diphenylmethyl)piperazin-1-yl]ethoxy)acetic Acid Dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.