Learn More
Campesterol-d7 (Mixture of Diastereomers), TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Campesterol-d7 (Mixture of Diastereomers) |
| CAS | 2483832-11-5 |
| Molecular Formula | C28 D7 H41 O |
| Quantity | 100 mg |
| Synonym | Ergost-5-en-25,26,26,26,27,27,27-d7-3-ol, (3β,24ξ)- (ACI), (3β,24ξ)-Ergost-5-en-25,26,26,26,27,27,27-d7-3-ol (ACI), (24ξ)-24-Methylcholest-5-en-3β-ol-25,26,26,26,27,27,27-d7, (3β,24ξ)-Ergost-5-en-3-ol-25,26,26,26,27,27,27-D7, 5-Cholesten-24(RS)-methyl-3β-ol-25,26,26,26,27,27,27-d7 |
| InChI Formula | InChI=1S/C28H48O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h9,18-20,22-26,29H,7-8,10-17H2,1-6H3/t19?,20-,22+,23+,24-,25+,26+,27+,28-/m1/s1/i1D3,2D3,18D |
| SMILES | [2H]C([2H])([2H])C([2H])(C(C)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C([2H])([2H])[2H] |
| IUPAC Name | (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6,7,7,7-tetradeuterio-5-methyl-6-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol |
| Molecular Weight (g/mol) | 407.7233 |
| Formula Weight | 407.4145 |