Learn More
Benzyl-2,3,4,5,6-D5 Alcohol, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Benzyl-2,3,4,5,6-D5 Alcohol |
| CAS | 68661-10-9 |
| Molecular Formula | C7 D5 H3 O |
| Quantity | 100 mg |
| Synonym | Benzene-D5-methanol, Benzene-2,3,4,5,6-D5-methanol, Benzylalcohol D5 (Phenyl D5), Benzene-d5-methanol (9CI), Benzyl-2,3,4,5,6-d5 alcohol (6CI), (2,3,4,5,6-Pentadeuteriophenyl)methanol, Benzene-2,3,4,5,6-d5-methanol (ACI) |
| InChI Formula | InChI=1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2/i1D,2D,3D,4D,5D |
| SMILES | [2H]c1c([2H])c([2H])c(CO)c([2H])c1[2H] |
| IUPAC Name | (2,3,4,5,6-pentadeuteriophenyl)methanol |
| Molecular Weight (g/mol) | 113.17 |
| Formula Weight | 113.0889 |