missing translation for 'onlineSavingsMsg'
Learn More

3-O-Methylfluorescein Phosphate Bis-Cyclohexylammonium Salt, TRC

Product Code. 30133554
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133554 100 mg 100mg
17920505 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133554 Supplier Toronto Research Chemicals Supplier No. TRCM227495100MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C21 H15 O8 P . 2 C6 H13 N
Molecular Weight (g/mol) 624.66
Synonym 3'-Hydroxy-6'-methoxy-fluoran Diphosphate Bis-Cyclohexylamine Salt
IUPAC Name cyclohexanamine;(6'-methoxy-3-oxo-spiro[isobenzofuran-1,9'-xanthene]-3'-yl) dihydrogen phosphate
SMILES COc1ccc2c(Oc3cc(OP(=O)(O)O)ccc3C24OC(=O)c5ccccc45)c1.NC6CCCCC6.NC7CCCCC7

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H15O8P.2C6H13N/c1-26-12-6-8-16-18(10-12)27-19-11-13(29-30(23,24)25)7-9-17(19)21(16)15-5-3-2-4-14(15)20(22)28-21;2*7-6-4-2-1-3-5-6/h2-11H,1H3,(H2,23,24,25);2*6H,1-5,7H2
Formula Weight 624.2601
Percent Purity >95%
Chemical Name or Material 3-O-Methylfluorescein Phosphate Bis-Cyclohexylammonium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.