missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, AISI O6, T31506, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Tool Steel, AISI O6, T31506, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Oxygen/Antimony/Calcium/Zirconium/Arsenic/Niobium/Titanium/Vanadium/Cobalt/Tungsten/Nitrogen/Tin/Aluminium/Phosphorus/Copper/Nickel/Chromium/Molybdenum/Manganese/Silicon/Carbon |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0001 %/Oxygen 0.0005 %/Antimony 0.001 %/Calcium 0.001 %/Zirconium 0.001 %/Arsenic 0.002 %/Niobium 0.002 %/Titanium 0.002 %/Vanadium 0.003 %/Cobalt 0.004 %/Tungsten 0.004 %/Nitrogen 0.008 %/Tin 0.008 %/Aluminium 0.01 %/Phosphorus 0.01 %/Copper 0.018 %/Nickel 0.024 %/Chromium 0.061 %/Molybdenum 0.24 %/Manganese 0.9 %/Silicon 0.92 %/Carbon 1.42 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction