missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cr/W/Mo, Alloy 230, N06230, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Nickel, Ni/Cr/W/Mo, Alloy 230, N06230, 100g
Specifications
Specifications
| Chemical Name or Material | Oxygen/Boron/Magnesium/Phosphorus/Lanthanum/Titanium/Vanadium/Niobium/Copper/Tantalum/Nitrogen/Carbon/Aluminium/Silicon/Cobalt/Manganese/Iron/Molybdenum/Tungsten/Chromium/Nickel/Zirconium |
| CAS | 7440-02-0 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Oxygen 0.0009 %/Boron 0.002 %/Magnesium 0.004 %/Phosphorus 0.005 %/Lanthanum 0.006 %/Titanium 0.008 %/Vanadium 0.013 %/Niobium 0.018 %/Copper 0.019 %/Tantalum 0.021 %/Nitrogen 0.05 %/Carbon 0.097 %/Aluminium 0.22 %/Silicon 0.38 %/Cobalt 0.4 %/Manganese 0.52 %/Iron 1.48 %/Molybdenum 2.34 %/Tungsten 13.45 %/Chromium 21.02 %/Nickel 59.8 %/Zirconium <0.01 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction