missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdB Labs AB 10 kDa DEAE-dextran for Transfection in Gene Therapy Studies
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
Description
- Product Overview: DEAE-dextran is a cationic, water-soluble polysaccharide modified with diethylaminoethyl groups. Known for its strong positive charge and high biocompatibility, DEAE-dextran is ideal for researchers and bioprocess engineers working with protein purification, nucleic acid complexation, or surface functionalization. Available across a wide molecular weight range to match different project needs.
- Key Features:
- Strong Cationic Properties: DEAE-modification provides consistent positive charge for robust binding to negatively charged biomolecules like proteins, DNA, and RNA.
- Flexible Molecular Weight Range: Offered from 4 kDa up to 2000 kDa, giving options for fine-tuning binding strength, diffusion, and separation in your application.
- High Solubility: Fully water-soluble powder allows rapid and easy preparation for chromatography, formulation, or complexation protocols.
- Batch-to-Batch Consistency: Tight control over molecular weight and functionalization ensures reliable results across experiments and production runs.
- Stable Storage: Dry powder format provides long shelf life without special storage conditions, helping to minimize lab downtime.
- Why Choose DEAE-dextran:
- For Protein Purification: DEAE-dextran is a proven choice for anion-exchange chromatography workflows, helping isolate or fractionate proteins under mild, biocompatible conditions.
- For Complexation and Binding Studies: Ideal for creating complexes with nucleic acids or proteins to study binding kinetics, surface adsorption, or biomolecular interactions.
- For Transfection Research: Historically used in mammalian cell DNA transfection experiments, DEAE-dextran still finds niche use where simple, non-viral gene delivery is studied.
- For Surface Modification: Researchers functionalizing surfaces (e.g., biosensors, biomaterials) benefit from DEAE-dextran's ability to introduce stable, positive surface charges.
- For Vaccine Adjuvant Studies: Studied as a component in veterinary vaccine formulations to enhance immune system targeting by improving antigen presentation.
- Typical Applications:
- Ion-exchange chromatography for proteins and nucleic acids
- Formation of electrostatic complexes for analytical and purification purposes
- Historical and research-based DNA transfection in mammalian cells
- Surface coating and modification in bioengineering projects
- Supportive studies in vaccine and adjuvant formulation development
- Technical Details:
- Molecular Weights: 4 kDa, 20 kDa, 40 kDa, 70 kDa, 150 kDa, 500 kDa, and 2000 kDa available.
- Charge Density: Approximately 0.1 to 0.2 DEAE groups per glucose unit (depending on the grade).
- Appearance: White to off-white dry powder, highly soluble in water.
- Storage and Handling:
- Store dry at room temperature in a sealed container to maintain stability.
- Protect from moisture to preserve full activity over long-term storage.
- Availability:
- Supplied as research-grade powder, suitable for laboratory use and pilot-scale process development.
- Available in small packs for R and D or bulk volumes for production environments.
Specifications
Specifications
| Cell Type | CHO |
| Format | Vial |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| Species | Human |
| Product Type | Transfection Reagent |
| Quantity | 100 g |
| Sterility | Non-sterile |
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido