missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, AISI 1018, G10180, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Carbon Steel, AISI 1018, G10180, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Calcium/Niobium/Zirconium/Titanium/Vanadium/Aluminium/Tungsten/Arsenic/Nitrogen/Cobalt/Tin/Phosphorus/Molybdenum/Sulfur/Nickel/Chromium/Carbon/Copper/Silicon/Manganese/Lead/Magnesium/Zinc/Oxygen/Tantalum/Antimony |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0003 %/Calcium 0.0006 %/Niobium 0.0011 %/Zirconium 0.0012 %/Titanium 0.0015 %/Vanadium 0.0016 %/Aluminium 0.003 %/Tungsten 0.005 %/Arsenic 0.0064 %/Nitrogen 0.0067 %/Cobalt 0.007 %/Tin 0.013 %/Phosphorus 0.019 %/Molybdenum 0.024 %/Sulfur 0.029 %/Nickel 0.087 %/Chromium 0.111 %/Carbon 0.17 %/Copper 0.27 %/Silicon 0.286 %/Manganese 0.82 %/Lead (0.0003) %/Magnesium (0.0005) %/Zinc (0.001) %/Oxygen (0.0017) %/Tantalum (0.002) %/Antimony (0.003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction