missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Beryllium Copper, CDA 172, C17200, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Copper, Beryllium Copper, CDA 172, C17200, 100g
Specifications
Specifications
| Chemical Name or Material | Antimony/Tin/Sulfur/Silver/Chromium/Cobalt/Manganese/Phosphorus/Aluminium/Silicon/Iron/Magnesium/Nickel/Beryllium/Bismuth/Selenium/Zinc/Arsenic/Cadmium/Lead/Copper |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Antimony 0.0007 %/Tin 0.0007 %/Sulfur 0.001 %/Silver 0.0011 %/Chromium 0.0017 %/Cobalt 0.0017 %/Manganese 0.0019 %/Phosphorus 0.0036 %/Aluminium 0.032 %/Silicon 0.045 %/Iron 0.071 %/Magnesium 0.112 %/Nickel 0.237 %/Beryllium 1.89 %/Bismuth (0.0004) %/Selenium (0.001) %/Zinc (0.003) %/Arsenic (0.004) %/Cadmium (0.004) %/Lead (0.004) %/Copper (97.6) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction