missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, 5Cr 1⁄2Mo, A182 Grade F5a, K42544, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Alloy Steel, 5Cr 1⁄2Mo, A182 Grade F5a, K42544, 100g
Specifications
Specifications
| Chemical Name or Material | Titanium/Niobium/Nitrogen/Arsenic/Tin/Aluminium/Tungsten/Phosphorus/Cobalt/Vanadium/Carbon/Copper/Nickel/Silicon/Manganese/Molybdenum/Chromium |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Titanium 0.0025 %/Niobium 0.004 %/Nitrogen 0.008 %/Arsenic 0.009 %/Tin 0.009 %/Aluminium 0.01 %/Tungsten 0.012 %/Phosphorus 0.014 %/Cobalt 0.015 %/Vanadium 0.017 %/Carbon 0.096 %/Copper 0.121 %/Nickel 0.148 %/Silicon 0.31 %/Manganese 0.408 %/Molybdenum 0.5 %/Chromium 4.34 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction