Learn More
Hamilton™ 1000 Series Standard PTFE Instrument Syringe
Setting the standard for quality and reliability.
161.00€ - 455.00€
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10132963
|
Hamilton™
5495-55 |
12.5 mL |
455.00€
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
11560222
|
Hamilton™
5495-45 |
2.5 mL |
161.00€
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Description
- Syringe life is exceptional when used in conjunction with Hamilton™ precision syringe drives
- Enables precision liquid handling
- Gastight for leak-free performance