Learn More
Hamilton™ 1000 Series Standard PTFE Instrument Syringe
Setting the standard for quality and reliability.
Brand:  Hamilton™ 5495-45
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11560222
  • 161.00€ / Each
Estimated Shipment: 21-06-2024
Description
  • Syringe life is exceptional when used in conjunction with Hamilton™ precision syringe drives
  • Enables precision liquid handling
  • Gastight for leak-free performance
Specifications
Specifications
2.5 mL