Learn More
Hamilton™ 1000 Series Gastight Instrument Syringes
With a precision-machined PTFE plunger tip which creates a leak-free seal.
135.00€ - 160.00€
Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15325104
View Documents Promotion Details
Hamilton™
208645
5 mL
135.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
15335104
View Documents Promotion Details
Hamilton™
208643
2.5 mL
160.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
Description
  • Tip wipes the interior of the syringe barrel free of sample
  • Useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze
  • Gas sterilizable