Learn More
Hamilton™ 1000 Series Gastight Instrument Syringes
With a precision-machined PTFE plunger tip which creates a leak-free seal.
135.00€ - 160.00€
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15325104
|
Hamilton™
208645 |
5 mL |
135.00€
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
15335104
|
Hamilton™
208643 |
2.5 mL |
160.00€
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
Description
- Tip wipes the interior of the syringe barrel free of sample
- Useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze
- Gas sterilizable