Learn More
Hamilton™ 1000 Series Gastight Instrument Syringes
With a precision-machined PTFE plunger tip which creates a leak-free seal.
Brand:  Hamilton™ 208645
Additional Details : Weight : 0.06000kg
 View more versions of this product
Product Code. 15325104
  • 135.00€ / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • Tip wipes the interior of the syringe barrel free of sample
  • Useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze
  • Gas sterilizable
Specifications
Specifications
5 mL