Learn More
Linker Industrietechnik™ Stainless Steel Test Sieve, 100 Dia. x 45 mm Height
Product Code. 11727166
Click to view available options
Pore Size:
0.25 μm
20 μm
25 μm
32 μm
36 μm
38 μm
40 μm
45 μm
50 μm
53 μm
56 μm
63 μm
71 μm
75 μm
80 μm
90 μm
1 mm
100 μm
106 μm
112 μm
125 μm
150 μm
1.6 mm
160 μm
1.8 mm
180 μm
2 mm
200 μm
212 μm
2.24 mm
2.5 mm
2.8 mm
280 μm
300 μm
355 μm
4 mm
400 μm
425 μm
500 μm
600 μm
630 μm
710 μm
8 mm
800 μm
850 μm
900 μm
10 mm
Unit Size:
Each
Product Code. 11727166
Brand: Linker Industrietechnik™ 551.040.200062
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Sifts particles in a wide range of pore sizes. These stainless-steel test sieves have an internal height of 45 mm and diameter of 100 mm.
  • Stainless-steel racks with an internal height of 45 mm
  • Diameter: 100 mm
  • Stainless-steel test sieve skin
  • Soft soldered seam
  • Identification number and certificate of compliance
  • Resistant up to temperatures of 170°C
Specifications
Quantity 1 Each
Pore Size 2 mm
Type Test Sieve
Certifications/Compliance Compliance 2.1 EN 10 204 and ISO 3310-1. Main range in accordance with ISO 565 (mesh apertures).
Material Stainless Steel
Height (Metric) 45mm
Diameter (Metric) Outer 100 mm
Mesh Material Stainless Steel