Learn More
Eppendorf™ 100/240VAC / 5VDC Power Supply
For Eppendorf Xplorer and electronic Eppendorf Repeater
Brand:  Eppendorf™ 4986603005
22.89 EUR valid until 2025-12-31
BEST PRICE on Promo! Use promo code "24145" to get your promotional price.
Product Code. 10084964
  • ONLINE EXCLUSIVE PRICE
    24.15€
    22.90€ / Each
    Save 1.25€
    5% Off
View all promotions
Only null left
In Stock
Description
  • Spare charging cable
  • For Eppendorf Xplorer and electronic Eppendorf Repeater
  • Includes charger stand