missing translation for 'onlineSavingsMsg'
Learn More

10-Phthalamido-1-decanol, TRC

Product Code. 30427969
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30427969 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30427969 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCP3824752.5G
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10-Phthalamido-1-decanol
CAS 161270-70-8
Molecular Formula C18H25NO3
Quantity 2.5 g
Synonym 2-(10-Hydroxydecyl)-1H-isoindole-1,3(2H)-dione
InChI Formula InChI=1S/C18H25NO3/c20-14-10-6-4-2-1-3-5-9-13-19-17(21)15-11-7-8-12-16(15)18(19)22/h7-8,11-12,20H,1-6,9-10,13-14H2
SMILES OCCCCCCCCCCN1C(=O)c2ccccc2C1=O
IUPAC Name 2-(10-hydroxydecyl)isoindole-1,3-dione
Molecular Weight (g/mol) 303.4
Formula Weight 303.1834
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.