missing translation for 'onlineSavingsMsg'
Learn More

10-Oxodecanoic Acid, TRC

Product Code. 30428591
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428591 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428591 Supplier Toronto Research Chemicals Supplier No. TRCO124310100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10-Oxodecanoic Acid
CAS 5578-80-3
Molecular Formula C10 H18 O3
Quantity 100 mg
InChI Formula InChI=1S/C10H18O3/c11-9-7-5-3-1-2-4-6-8-10(12)13/h9H,1-8H2,(H,12,13)
SMILES OC(=O)CCCCCCCCC=O
IUPAC Name 10-oxodecanoic acid
Molecular Weight (g/mol) 186.248
Formula Weight 186.126
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.