Learn More
10-Carboxylinalool, TRC
Product Code. 30240879
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240879 25 mg 25mg
30240719 5 mg 5mg
30240715 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30240879 Supplier Toronto Research Chemicals Supplier No. TRCC24096525MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28420-25-9
Molecular Formula C10 H16 O3
Molecular Weight (g/mol) 184.232
Synonym 6-Hydroxy-2,6-dimethyl-2,7-octadienoic Acid;
IUPAC Name (2E)-6-hydroxy-2,6-dimethylocta-2,7-dienoic acid
SMILES C\C(=C/CCC(C)(O)C=C)\C(=O)O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C10H16O3/c1-4-10(3,13)7-5-6-8(2)9(11)12/h4,6,13H,1,5,7H2,2-3H3,(H,11,12)/b8-6+
Formula Weight 184.11
Chemical Name or Material 10-Carboxylinalool