Learn More
Fisherbrand™ -10 to +330 Lo Tox™ Total Immersion Laboratory Thermometer
Shop All Fisherbrand ProductsDescription
- Quality and legibility make these an ideal substitute for mercury thermometers
- Fitted with Brannan non roll end cap which has ring to allow the thermometer to be suspended
Specifications
Specifications
| Length (Metric) | 305 mm |
| Thermometer Fill | Brown Organic Filling |
| Product Type | Total Immersion Laboratory Thermometer |
| Max. Temperature (Metric) | 330°C |
| Min. Temperature (Metric) | -10°C |
| Temperature Range (Metric) | -10°C to +330° C |
| Accuracy | ±2°C at 90°C |
| Subdivision | 2° |