missing translation for 'onlineSavingsMsg'
Learn More

10,12-Pentacosadiynoic acid, 98+%

Product Code. 11337336
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11337336

Brand: Thermo Scientific Alfa Aesar L09313.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 66990-32-7 | C25H42O2 | 374.61 g/mol

10,12-Pentacosadiynoic acid is used in the preparation of colorimetric and fluorescence sensors for cationic surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10,12-Pentacosadiynoic acid is used in the preparation of colorimetric and fluorescence sensors for cationic surfactants.

Solubility
Slightly soluble in water.

Notes
Incompatible with reducing agents and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66990-32-7
Molecular Formula C25H42O2
Molecular Weight (g/mol) 374.61
MDL Number MFCD00041684
InChI Key ZPUDRBWHCWYMQS-UHFFFAOYSA-N
Synonym 10,12-pentacosadiynoic acid, 10,12-pentacosadiynoicacid, acmc-209nx9, 10-12-pentacosadiynoic acid, 10,12-pentacosadiynoic acid hplc
PubChem CID 538433
IUPAC Name pentacosa-10,12-diynoic acid
SMILES CCCCCCCCCCCCC#CC#CCCCCCCCCC(O)=O

Specifications

Melting Point 62°C to 65°C
Quantity 25 g
Beilstein 4189973
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 374.61
Percent Purity ≥98%
Chemical Name or Material 10,12-Pentacosadiynoic acid

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis