Learn More
10,11-Dihydro-10-hydroxy Carbamazepine, TRC
Specifications
Specifications
| Chemical Name or Material | 10,11-Dihydro-10-Hydroxycarbamazepine |
| CAS | 29331-92-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 H14 N2 O2 |
| Quantity | 100 mg |
| Synonym | 10-Hydroxy-10,11-dihydrocarbamazepine, 10,11-Dihydro-10-hydroxy-5H-dibenz[b,f]azepine-5-carboxamide, 10,11-Dihydro-10-hydroxycarbamazepine, 10-Hydroxy-10,11-dihydrocarbamezepine, BIA 2-005, GP 47779, Licarbazepine |
| InChI Formula | InChI=1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19) |
| SMILES | NC(=O)N1c2ccccc2CC(O)c3ccccc13 |
| IUPAC Name | 5-hydroxy-5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide |
| Molecular Weight (g/mol) | 254.28 |
| Show More |