Learn More
1-(tert-Butylamino)-3-(1-naphthyloxy)-2-propanol, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(tert-Butylamino)-3-(1-naphthyloxy)-2-propanol |
| CAS | 2116-22-5 |
| Molecular Formula | C17H23NO2 |
| Quantity | 250 mg |
| Synonym | 1-[(1,1-Dimethylethyl)amino]-3-(1-naphthalenyloxy)-2-propanol, Nadolol EP Impurity F, (RS)-2'-Methylpropranolol, (±)-2'-Methylpropranolol |
| InChI Formula | InChI=1S/C17H23NO2/c1-17(2,3)18-11-14(19)12-20-16-10-6-8-13-7-4-5-9-15(13)16/h4-10,14,18-19H,11-12H2,1-3H3 |
| SMILES | CC(C)(C)NCC(O)COc1cccc2ccccc12 |
| IUPAC Name | 1-(tert-butylamino)-3-naphthalen-1-yloxypropan-2-ol |
| Molecular Weight (g/mol) | 273.37 |
| Formula Weight | 273.1729 |