Learn More
1-t-Boc-piperidine-4-spiro-5’-hydantoin, TRC
Specifications
Specifications
| Chemical Name or Material | 1-t-Boc-piperidine-4-spiro-5'-hydantoin |
| CAS | 183673-70-3 |
| Molecular Formula | C12 H19 N3 O4 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C12H19N3O4/c1-11(2,3)19-10(18)15-6-4-12(5-7-15)8(16)13-9(17)14-12/h4-7H2,1-3H3,(H2,13,14,16,17) |
| SMILES | CC(C)(C)OC(=O)N1CCC2(CC1)NC(=O)NC2=O |
| IUPAC Name | tert-butyl 2,4-dioxo-1,3,8-triazaspiro[4.5]decane-8-carboxylate |
| Molecular Weight (g/mol) | 269.3 |
| Formula Weight | 269.1376 |