missing translation for 'onlineSavingsMsg'
Learn More

1-Stearoyl-rac-glycerol, TRC

Product Code. 17967534
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17967534 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17967534 Supplier Toronto Research Chemicals Supplier No. TRCS686560100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 123-94-4
Molecular Formula C21 H42 O4
Quantity 100 mg
Synonym Glyceryl 1-Monostearate, Stearin, 1-mono- (6CI, 7CI, 8CI), (±)-2,3-Dihydroxypropyl octadecanoate, 1-Glyceryl stearate, 1-Monooctadecanoylglycerol, 1-Monostearin, 1-Monostearoyl-rac-glycerol, 1-monostearoyl-rac-glycerol, 1-Monostearoylglycerol, 1-Stearoyl-rac-glycerol, 2,3-Dihydroxypropyl octadecanoate, 2,3-Dihydroxypropyl stearate, 3-Stearoyloxy-1,2-propanediol, Aldo 33, Aldo 75, Aldo MSD, Aldo MSLG, AS 8 (surfactant), Emerest 2407, Glycerin 1-monostearate, Glycerin 1-stearate, Glycerol 1-monostearate, Glycerol 1-stearate, Glycerol α-monostearate, Glyceryl 1-monostearate, Glyceryl 1-octadecanoate, NSC 3875, Sandin EU, Stearic acid 1-monoglyceride, Stearic acid α-monoglyceride, Tegin 55G, α-Monostearin
InChI Formula InChI=1S/C21H42O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h20,22-23H,2-19H2,1H3
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO
IUPAC Name 2,3-dihydroxypropyl octadecanoate
Molecular Weight (g/mol) 358.56
Formula Weight 358.3083
Chemical Name or Material 1-Stearoyl-rac-glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.