Learn More
(1’R,2R)-2-(1’,2’-Dihydroxyethyl)-6-fluorochromane, TRC
Specifications
Specifications
| Chemical Name or Material | (1'R,2R)-2-(1',2'-Dihydroxyethyl)-6-fluorochromane |
| CAS | 303176-45-6 |
| Molecular Formula | C11 H13 F O3 |
| Quantity | 100 mg |
| Synonym | (1R)-1-[(2R)-6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-yl]-1,2-ethanediol |
| InChI Formula | InChI=1S/C11H13FO3/c12-8-2-4-10-7(5-8)1-3-11(15-10)9(14)6-13/h2,4-5,9,11,13-14H,1,3,6H2/t9-,11-/m1/s1 |
| SMILES | OC[C@@H](O)[C@H]1CCc2cc(F)ccc2O1 |
| IUPAC Name | (1R)-1-[(2R)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]ethane-1,2-diol |
| Molecular Weight (g/mol) | 212.218 |
| Formula Weight | 212.085 |