Learn More
1-Pyrenemethanol, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Pyrenemethanol |
| CAS | 24463-15-8 |
| Molecular Formula | C17 H12 O |
| Quantity | 500 mg |
| Synonym | 1-Pyrenemethanol, 1-(1-Hydroxymethyl)pyrene, 1-Hydroxymethylpyrene |
| InChI Formula | InChI=1S/C17H12O/c18-10-14-7-6-13-5-4-11-2-1-3-12-8-9-15(14)17(13)16(11)12/h1-9,18H,10H2 |
| SMILES | OCc1ccc2ccc3cccc4ccc1c2c34 |
| IUPAC Name | pyren-1-ylmethanol |
| Molecular Weight (g/mol) | 232.28 |
| Formula Weight | 232.0888 |