Learn More
1-Propanethiol, TRC
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Specifications
Specifications
| Chemical Name or Material | 1-Propanethiol |
| CAS | 107-03-9 |
| Molecular Formula | C3 H8 S |
| Quantity | 50 mL |
| Synonym | 1-Mercaptopropane, 1-Propyl mercaptan, 1-Propylthiol, Propyl mercaptan, Propylthiol, n-Propanethiol, n-Propyl mercaptan, n-Propylthiol |
| InChI Formula | InChI=1S/C3H8S/c1-2-3-4/h4H,2-3H2,1H3 |
| SMILES | CCCS |
| IUPAC Name | propane-1-thiol |
| Molecular Weight (g/mol) | 76.16 |
| Formula Weight | 76.0347 |