missing translation for 'onlineSavingsMsg'
Learn More

1-Piperonylpiperazine, 97%, Thermo Scientific™

Product Code. p-7046666
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318366 5 g 5g
11328366 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318366 Supplier Thermo Scientific Alfa Aesar Supplier No. L15625.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32231-06-4
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD00005834
InChI Key NBOOZXVYXHATOW-UHFFFAOYSA-N
Synonym 1-piperonylpiperazine, 1-3,4-methylenedioxybenzyl piperazine, 1-benzo d 1,3 dioxol-5-ylmethyl piperazine, 1-1,3-benzodioxol-5-ylmethyl piperazine, 1-1,3-benzodioxol-5-yl-methyl piperazine, 1-benzo 1,3 dioxol-5-ylmethyl-piperazine, piperazine, 1-1,3-benzodioxol-5-ylmethyl, 1-3,4-methylendioxybenzyl piperazine, piperazine, 1-3,4-methylenedioxybenzyl, 5-piperazinylmethyl-2h-benzo d 1,3-dioxolene
PubChem CID 94426
IUPAC Name 1-(1,3-benzodioxol-5-ylmethyl)piperazine
SMILES C1CN(CCN1)CC2=CC3=C(C=C2)OCO3

Specifications

Melting Point 40°C to 45°C
Boiling Point 147°C to 149°C (2 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3259
Beilstein 885038
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material 1-Piperonylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.