missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpentan-2-amine, TRC

Product Code. 30335964
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335964 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335964 Supplier Toronto Research Chemicals Supplier No. TRCB522708250MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Phenylpentan-2-amine
CAS 63951-01-9
Molecular Formula C11H17N
Quantity 250 mg
Synonym alpha-Propylbenzeneethanamine, alpha-Propylphenethylamine
InChI Formula InChI=1S/C11H17N/c1-2-6-11(12)9-10-7-4-3-5-8-10/h3-5,7-8,11H,2,6,9,12H2,1H3
SMILES CCCC(N)Cc1ccccc1
IUPAC Name 1-phenylpentan-2-amine
Molecular Weight (g/mol) 163.25
Formula Weight 163.1361
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.