Learn More
1-Phenylbutan-1-amine, TRC
Product Code. 30427760
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427760 50 mg 50mg
30418736 500 mg 500mg
30415208 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30427760 Supplier Toronto Research Chemicals Supplier No. TRCP32100050MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2941-19-7
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.23
IUPAC Name 1-phenylbutan-1-amine
SMILES CCCC(N)c1ccccc1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C10H15N/c1-2-6-10(11)9-7-4-3-5-8-9/h3-5,7-8,10H,2,6,11H2,1H3
Formula Weight 149.1204
Chemical Name or Material 1-Phenylbutan-1-amine