Learn More
1-Phenyl-3-azabicyclo[3.1.0]hexan-2-one, TRC
Product Code. 30427608
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427608 1 g 1g
30417281 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30427608 Supplier Toronto Research Chemicals Supplier No. TRCP3208101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 153275-06-0
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.21
Synonym 1-Phenyl-3-aza-bicyclo[3.1.0]hexan-2-one
IUPAC Name 1-phenyl-3-azabicyclo[3.1.0]hexan-2-one
SMILES O=C1NCC2CC12c3ccccc3
Specifications
Quantity 1 g
InChI Formula InChI=1S/C11H11NO/c13-10-11(6-9(11)7-12-10)8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,12,13)
Formula Weight 173.0841
Chemical Name or Material 1-Phenyl-3-azabicyclo[3.1.0]hexan-2-one