missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-1-heptanone, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 1-Phenyl-1-heptanone |
| CAS | 1671-75-6 |
| Molecular Formula | C13H18O |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C13H18O/c1-2-3-4-8-11-13(14)12-9-6-5-7-10-12/h5-7,9-10H,2-4,8,11H2,1H3 |
| SMILES | CCCCCCC(=O)c1ccccc1 |
| IUPAC Name | 1-phenylheptan-1-one |
| Molecular Weight (g/mol) | 190.28 |
| Formula Weight | 190.1358 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction