Learn More
1-Phenyl-1,2,3,4-tetrahydronaphthalene, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Phenyl-1,2,3,4-tetrahydronaphthalene |
| CAS | 3018-20-0 |
| Molecular Formula | C16 H16 |
| Quantity | 10 mg |
| Synonym | 1,2,3,4-Tetrahydro-1-phenylnaphthalene, 1-Phenyltetralin. |
| InChI Formula | InChI=1S/C16H16/c1-2-7-13(8-3-1)16-12-6-10-14-9-4-5-11-15(14)16/h1-5,7-9,11,16H,6,10,12H2 |
| SMILES | C1CC(c2ccccc2)c3ccccc3C1 |
| IUPAC Name | 1-phenyl-1,2,3,4-tetrahydronaphthalene |
| Molecular Weight (g/mol) | 208.298 |
| Formula Weight | 208.125 |