Learn More
1-Pentadecyne, 98%
Product Code. 11397735
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397735 2 g 2g
11307745 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11397735 Supplier Thermo Scientific Alfa Aesar Supplier No. L02540.04
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 765-13-9 | C15H28 | 208.39 g/mol
1-Pentadecyne is used in pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Pentadecyne is used in pharmaceutical research.
Solubility
Not miscible in water.
Notes
Store under inert gas. Air sensitive. Avoid excessive heat and light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 765-13-9
Molecular Formula C15H28
Molecular Weight (g/mol) 208.39
MDL Number MFCD00015081
InChI Key DONJGKADZJEXRJ-UHFFFAOYSA-N
Synonym 1-pentadecyne, 1-penatdecyne, pentadec-1-in, tridecylacetylene, 1-pentadecyne 1g, 1-pentadecyne,
PubChem CID 69825
IUPAC Name pentadec-1-yne
SMILES CCCCCCCCCCCCCC#C
Specifications
Melting Point 10°C
Density 0.793
Boiling Point 129°C to 131°C (10 mmHg)
Flash Point >90°C (194°F)
Refractive Index 1.442
Quantity 2 g
Beilstein 1751009
Solubility Information Not miscible in water.
Formula Weight 208.39
Percent Purity 98%
Chemical Name or Material 1-Pentadecyne