missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Pregnen-3b,20b-diol-17,20,21,21,21-d5 3-Acetate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (3 beta,20 R)-Pregn-5-ene-3,20-diol-17,20,21,21,21-D5 3-Acetate |
| Molecular Formula | C23 D5 H31 O3 |
| Quantity | 0.01 g |
| Synonym | 3β-Acetoxy-5-pregnen-20β-ol-17,20,21,21,21-d5, Pregn-5-ene-3β,20β-diol-17,20,21,21,21-d5 3-acetate, (3β,20 R)-Pregn-5-ene-3,20-diol-17,20,21,21,21-D5 3-Acetate, 3β-Acetoxy-20β-dihydropregnenolone-17,20,21,21,21-d5, 3β-Acetoxy-20β-hydroxypregn-5-ene-17,20,21,21,21-d5, delta;5-Pregnene-3β,20β-diol-17,20,21,21,21-d5 3-acetate |
| InChI Formula | InChI=1 S/C23H36O3/c1-14(24)19-7-8-20-18-6-5-16-13-17(26-15(2)25)9-11-22(16,3)21(18)10-12-23(19,20)4/h5,14,17-21,24 H,6-13H2,1-4H3/t14-,17+,18+,19-,20+,21+,22+,23-/m1/s1/i1D3,14 D,19 D |
| SMILES | [2 H]C([2 H])([2 H])[C@@]([2 H])(O)[C@@]1([2 H])CC[C@H]2 [C@@H]3 CC=C4C[C@H](CC[C@]4(C)[C@H]3 CC[C@]12 C)OC(=O)C |
| IUPAC Name | [(3 S,8 S,9 S,10 R,13 S,14 S,17 S)-17-deuterio-10,13-dimethyl-17-[(1 R)-1,2,2,2-tetradeuterio-1-hydroxyethyl]-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-yl] acetate |
| Molecular Weight (g/mol) | 365.561 |
| Formula Weight | 365.2978 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction