Learn More
1-Naphthyl b-D-glucopyranoside, TRC
Product Code. 30476547
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476547 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30476547 Supplier Toronto Research Chemicals Supplier No. TRCN3720602.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Naphtyl-beta-D-Glucopyranoside
CAS 19939-82-3
Molecular Formula C16 H18 O6
Quantity 2.5 g
Synonym 1-Naphthol glucoside, α-Naphthol glucoside
InChI Formula InChI=1S/C16H18O6/c17-8-12-13(18)14(19)15(20)16(22-12)21-11-7-3-5-9-4-1-2-6-10(9)11/h1-7,12-20H,8H2/t12-,13-,14+,15-,16-/m1/s1
SMILES OC[C@H]1O[C@@H](Oc2cccc3ccccc23)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(1-naphthyloxy)tetrahydropyran-3,4,5-triol
Molecular Weight (g/mol) 306.31
Formula Weight 306.1103