Learn More
1-N-Methyl Lenalidomide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(4-Amino-1-oxo-isoindolin-2-yl)-1-methyl-piperidine-2,6-dione |
| CAS | 2133816-23-4 |
| Molecular Formula | C14 H15 N3 O3 |
| Quantity | 25 mg |
| Synonym | 2,6-Piperidinedione','3-(4-amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)-1-methyl- (ACI), 3-(4-Amino-1-oxo-isoindolin-2-yl)-1-methyl-piperidine-2,6-dione |
| InChI Formula | InChI=1S/C14H15N3O3/c1-16-12(18)6-5-11(14(16)20)17-7-9-8(13(17)19)3-2-4-10(9)15/h2-4,11H,5-7,15H2,1H3 |
| SMILES | CN1C(=O)CCC(N2Cc3c(N)cccc3C2=O)C1=O |
| IUPAC Name | 3-(7-amino-3-oxo-1H-isoindol-2-yl)-1-methylpiperidine-2,6-dione |
| Molecular Weight (g/mol) | 273.287 |
| Formula Weight | 273.111 |