Learn More
1-Methylpiperidine-3-methanol, 95%, Thermo Scientific™
Product Code. 11460668
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460668 50 g 50g
11470668 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11460668 Supplier Thermo Scientific Alfa Aesar Supplier No. B25197.18
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7583-53-1
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00006497
InChI Key UGXQXVDTGJCQHR-UHFFFAOYSA-N
Synonym 1-methyl-3-piperidinemethanol, 1-methylpiperidin-3-yl methanol, 1-methylpiperidine-3-methanol, 3-piperidinemethanol, 1-methyl, 3-hydroxymethyl-1-methylpiperidine, 1-methyl-3-piperidyl methanol, 1-methyl-3-hydroxymethylpiperidine, 1-methyl-piperidin-3-yl-methanol, 3-piperidinemethanol,1-methyl-,--9ci
PubChem CID 97998
IUPAC Name (1-methylpiperidin-3-yl)methanol
SMILES CN1CCCC(C1)CO
Specifications
Density 1.013
Boiling Point 140°C to 145°C
Flash Point 94°C (201°F)
Refractive Index 1.48
Quantity 50 g
Beilstein 103713
Formula Weight 129.2
Percent Purity 98%
Chemical Name or Material 1-Methylpiperidine-3-methanol