Learn More
1-Methylimidazole-d6, TRC
Product Code. 30481990
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481990 10 mg 10mg
30472935 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30481990 Supplier Toronto Research Chemicals Supplier No. TRCM31298910MG
 Restricted Item
Estimated Shipment: 12-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 285978-27-0
Molecular Formula C4 D6 N2
Molecular Weight (g/mol) 88.1408
Synonym 1H-Imidazole-2,4,5-d3, 1-(methyl-d3)- (9CI, ACI), 1-(Methyl-d3)-1H-imidazole-2,4,5-d3 (ACI), 1-(Methyl-d3)-1H-imidazole-2,4,5-d3, N-Methylimidazole-d6, 1-Methyl-1H-imidazole-D6, 1-Methylimidazole-d6
IUPAC Name 2,4,5-trideuterio-1-(trideuteriomethyl)imidazole
SMILES [2H]c1nc([2H])n(c1[2H])C([2H])([2H])[2H]
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C4H6N2/c1-6-3-2-5-4-6/h2-4H,1H3/i1D3,2D,3D,4D
Formula Weight 88.0908
Chemical Name or Material 1-Methylimidazole-d6