missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylimidazole-d3 (ring-d3), CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Methylimidazole-d3 (ring-d3) |
| CAS | 4166-68-1 |
| Molecular Formula | C4 D3 H3 N2 |
| Quantity | 0.25 g |
| Synonym | 1 H-Imidazole-2,4,5-d3, 1-methyl- (9 CI, ACI), 1-Methyl-1 H-imidazole-2,4,5-d3 (ACI), Imidazole-2,4,5-d3, 1-methyl- (7 CI, 8 CI), 1-Methylimidazole-2,4,5-d3, N-Methylimidazole-d3, 1-Methyl-1 H-imidazole-2,4,5-D3, 1-Methyl-1 H-imidazole-2,4,5-D3, 1-Methyl-1 H-imidazole-2,4,5-D3 |
| InChI Formula | InChI=1 S/C4H6N2/c1-6-3-2-5-4-6/h2-4 H,1H3/i2D,3 D,4 D |
| SMILES | [2 H]c1nc([2 H])n(C)c1[2 H] |
| IUPAC Name | 2,4,5-trideuterio-1-methylimidazole |
| Molecular Weight (g/mol) | 85.122 |
| Formula Weight | 85.072 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction