missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclohexanecarboxylic Acid-d10 Methyl Ester, TRC

Product Code. 30481528
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481528 5 mg 5mg
30478723 1 mg 1mg
30470809 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30481528 Supplier Toronto Research Chemicals Supplier No. TRCM2948375MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Methylcyclohexanecarboxylic Acid-d10 Methyl Ester
Molecular Formula C9 D10 H6 O2
Quantity 5 mg
Synonym Methyl 1-Methyl-1-cyclohexanecarboxylate-d10, Methyl 1-Methylcyclohexanecarboxylate-d10
InChI Formula InChI=1S/C9H16O2/c1-9(8(10)11-2)6-4-3-5-7-9/h3-7H2,1-2H3/i3D2,4D2,5D2,6D2,7D2
SMILES [2H]C1([2H])C([2H])([2H])C([2H])([2H])C(C)(C(=O)OC)C([2H])([2H])C1([2H])[2H]
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6-decadeuterio-1-methylcyclohexane-1-carboxylate
Molecular Weight (g/mol) 166.284
Formula Weight 166.178
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.