Learn More
1-Methylazepan-2-one, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2556-73-2 |
| Molecular Formula | C7H13NO |
| MDL Number | MFCD00003263 |
| Quantity | 5 g |
| SMILES | O=C1N(C)CCCCC1 |
| Molecular Weight (g/mol) | 127.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-Methylazepan-2-one |