Learn More
1-Methyl-5-nitroimidazole, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Methyl-5-nitroimidazole |
| CAS | 3034-42-2 |
| Molecular Formula | C4H5N3O2 |
| Quantity | 1 g |
| Synonym | 1-Methyl-5-nitro-1H-imidazole |
| InChI Formula | InChI=1S/C4H5N3O2/c1-6-3-5-2-4(6)7(8)9/h2-3H,1H3 |
| SMILES | Cn1cncc1[N+](=O)[O-] |
| IUPAC Name | 1-methyl-5-nitroimidazole |
| Molecular Weight (g/mol) | 127.1 |
| Formula Weight | 127.0382 |