Learn More
1-Methyl-4-(phenylsulfonyl)benzene, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 640-57-3 |
| Molecular Formula | C13H12O2S |
| MDL Number | MFCD00025038 |
| Quantity | 25 g |
| SMILES | O=S(C1=CC=C(C)C=C1)(C2=CC=CC=C2)=O |
| Molecular Weight (g/mol) | 232.30 |
| Percent Purity | 95% |
| Chemical Name or Material | 1-Methyl-4-(phenylsulfonyl)benzene |