Learn More
1-Methyl-3-phenylpropylamine, TRC
Product Code. 30135432
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135432 25 g 25g
30137069 100 g 100g
18020950 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135432 Supplier Toronto Research Chemicals Supplier No. TRCM32547025G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22374-89-6
Molecular Formula C10 H15 N
Molecular Weight (g/mol) 149.23
Synonym α-Methylbenzenepropanamine, (3-Aminobutyl)benzene, (RS)-1-Methyl-3-
IUPAC Name 4-phenylbutan-2-amine
SMILES CC(N)CCc1ccccc1
Specifications
Quantity 25 g
InChI Formula InChI=1S/C10H15N/c1-9(11)7-8-10-5-3-2-4-6-10/h2-6,9H,7-8,11H2,1H3
Formula Weight 149.1204
Chemical Name or Material 1-Methyl-3-Phenylpropylamine