Learn More
1-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 10328-92-4 |
| Molecular Formula | C9H7NO3 |
| MDL Number | MFCD00006815 |
| Quantity | 25 g |
| SMILES | O=C(N(C)C1=CC=CC=C12)OC2=O |
| Molecular Weight (g/mol) | 177.16 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione |