missing translation for 'onlineSavingsMsg'
Få mere at vide

(1-Methyl-1,2-ethanediyl)bis-Carbamodithioic Acid Dimethyl Ester, TRC

Produktkode 30134408
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 mg
5 mg
10 mg
Förpackningsstorlek
10mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity unitSize
30134408 10 mg 10mg
17953715 5 mg 5mg
17921005 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 30134408 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM62120010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 80555-60-8
Molecular Formula C7H14N2S4
Molecular Weight (g/mol) 254.46
Synonym S,S'-Dimethylxanthogenpropylenebisdithiocarbamate, Dimethylxanthogenpropylenebisdithiocarbamate: S,S'-Dimethylxanthogen-1,2-propylenebisdithiocarbamate
IUPAC Name methyl N-[1-(methylsulfanylcarbothioylamino)propan-2-yl]carbamodithioate
SMILES CSC(=S)NCC(C)NC(=S)SC

Tekniske data

Quantity 10 mg
InChI Formula InChI=1S/C7H14N2S4/c1-5(9-7(11)13-3)4-8-6(10)12-2/h5H,4H2,1-3H3,(H,8,10)(H,9,11)
Formula Weight 254.004
Percent Purity >95%
Chemical Name or Material (1-Methyl-1,2-ethanediyl)bis-Carbamodithioic Acid Dimethyl Ester
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.